Learn More
Chemglass Life Sciences Chemglass AccuPet Pro Single-Channel Variable Micropipette, 20-200 μL
SDP
Catalog No. NC1884659
Encompass_Preferred
The AccuPet Pro single-channel variable micropipette delivers accurate, repeatable liquid handling across a 20-200 μL range for routine and research laboratory use. It combines an easy-to-read volume indicator, ergonomic design, and an autoclavable lower portion for improved usability and maintenance.
  • Volume range: 20-200 μL.
  • Resolution: 1 μL.
  • Accuracy: ±2% at 20 μL; ±0.6% at 200 μL.
  • Precision (imprecision): ±0.8% at 20 μL; ±0.15% at 200 μL.
  • Single-channel air-displacement design.
  • Continuously adjustable volume with easy-to-read volume indicator.
  • Ergonomic handle with rubber grip for reduced fatigue.
  • Tip ejector and universal tip fitting shafts.
  • Autoclavable lower portion for sterilization.
  • Sold individually.
Catalog No. NC1884659 Supplier Chemglass Life Sciences Supplier No. CLS3489E06
The AccuPet Pro single-channel variable micropipette delivers accurate, repeatable liquid handling across a 20-200 μL range for routine and research laboratory use. It combines an easy-to-read volume indicator, ergonomic design, and an autoclavable lower portion for improved usability and maintenance.
  • Volume range: 20-200 μL.
  • Resolution: 1 μL.
  • Accuracy: ±2% at 20 μL; ±0.6% at 200 μL.
  • Precision (imprecision): ±0.8% at 20 μL; ±0.15% at 200 μL.
  • Single-channel air-displacement design.
  • Continuously adjustable volume with easy-to-read volume indicator.
  • Ergonomic handle with rubber grip for reduced fatigue.
  • Tip ejector and universal tip fitting shafts.
  • Autoclavable lower portion for sterilization.
  • Sold individually.