Learn More
Chemglass Life Sciences AccuPet Pro Air Displacement 0.5-10µL, 12-Channel Pipette
SDP
Catalog No. 5000980686
Encompass_Preferred
Ejector. Autoclavable Lower Portion. Easy to view volume indicator. Ergonomic handle design. The AccuPet Pro Multi comes in a variety of 8- and 12-channel pipettes (0.5-10µl, 5-50µl, 20-300µl.) This digital pipette is ergonomically designed and allows the user to pipette more comfortably and efficiently. The AccuPet Pro is designed with features such as the EasyCal technique, the DirectVue Volume Indicator, and an Ergonomic Handle Design in order to optimize pipetting.
Catalog No. 50-009-80686 Supplier Chemglass Life Sciences Supplier No. CLS3489M06
Ejector. Autoclavable Lower Portion. Easy to view volume indicator. Ergonomic handle design. The AccuPet Pro Multi comes in a variety of 8- and 12-channel pipettes (0.5-10µl, 5-50µl, 20-300µl.) This digital pipette is ergonomically designed and allows the user to pipette more comfortably and efficiently. The AccuPet Pro is designed with features such as the EasyCal technique, the DirectVue Volume Indicator, and an Ergonomic Handle Design in order to optimize pipetting.