Learn More
Chemglass Life Sciences AccuPet Pro Micropipette, 0.1-2.5µL, Single Channel Variable Pipettors
SDP
Catalog No. 5000980670
Encompass_Preferred
Ergonomic handle designed for comfort. Research grade, digital accuracy. Easy to view volume indicator. Continuously adjustable. Tip ejectors. Autoclavable lower portion. AccuPet Pro Micropipettes are the standard for microliter measurements. This line of digital pipettes is ergonomically designed and continuously adjustable. The AccuPet Pro is crafted to incorporate attractive form with practical function at an affordable price. The updated features allow for more comfortable pipetting and produce more precise results. These features include: the DirectVue Volume Indicator, a SuperLite Ergonomic Handle Design with a Rubber Grip, and Universal Tip Fitting Shafts. Please Note: Accuracy and precision value given at maximum volume of pipettor. Please Note: No Tip Ejector on the CLS-3489E-09.
Catalog No. 50-009-80670 Supplier Chemglass Life Sciences Supplier No. CLS3489E01
Ergonomic handle designed for comfort. Research grade, digital accuracy. Easy to view volume indicator. Continuously adjustable. Tip ejectors. Autoclavable lower portion. AccuPet Pro Micropipettes are the standard for microliter measurements. This line of digital pipettes is ergonomically designed and continuously adjustable. The AccuPet Pro is crafted to incorporate attractive form with practical function at an affordable price. The updated features allow for more comfortable pipetting and produce more precise results. These features include: the DirectVue Volume Indicator, a SuperLite Ergonomic Handle Design with a Rubber Grip, and Universal Tip Fitting Shafts. Please Note: Accuracy and precision value given at maximum volume of pipettor. Please Note: No Tip Ejector on the CLS-3489E-09.