Learn More
Chemglass Life Sciences Chemglass AccuPet Pro single-channel adjustable micropipette, 0.5-10 μL
SDP
Catalog No. NC3356232
Encompass_Preferred
Single-channel, research-grade adjustable air-displacement micropipette for accurate microliter delivery in molecular biology and analytical workflows. Designed for comfort and repeatability with an easy-to-read volume indicator and an autoclavable lower portion.
  • Ergonomic handle designed for comfort.
  • Easy-to-view direct volume indicator for quick setting verification.
  • Continuously adjustable volume with 0.1 μL increments.
  • High accuracy and precision at low and maximum test volumes.
  • Autoclavable lower portion for routine sterilization.
  • Universal tip fitting for compatibility with common pipette tips.
Catalog No. NC3356232 Supplier Chemglass Life Sciences Supplier No. CLS3489E02
Single-channel, research-grade adjustable air-displacement micropipette for accurate microliter delivery in molecular biology and analytical workflows. Designed for comfort and repeatability with an easy-to-read volume indicator and an autoclavable lower portion.
  • Ergonomic handle designed for comfort.
  • Easy-to-view direct volume indicator for quick setting verification.
  • Continuously adjustable volume with 0.1 μL increments.
  • High accuracy and precision at low and maximum test volumes.
  • Autoclavable lower portion for routine sterilization.
  • Universal tip fitting for compatibility with common pipette tips.