Learn More
Perkin Elmer US LLC AccuPik Needle Kit - Qty. 3
Catalog No. 502113382
Encompass_Preferred
The Accupik Accessory Kit works with the TGA 8000 with Autosampler units. Attached to the underside of the Balance Mounting Plate, the AccuPik pierces a crimped sample pan just before the pan enters the furnace, protecting sample against evaporation and volatilization. This accessory includes the puncture device and three replacement needles.
Catalog No. 50-211-3382 Supplier Perkin Elmer US LLC Supplier No. N5370554
The Accupik Accessory Kit works with the TGA 8000 with Autosampler units. Attached to the underside of the Balance Mounting Plate, the AccuPik pierces a crimped sample pan just before the pan enters the furnace, protecting sample against evaporation and volatilization. This accessory includes the puncture device and three replacement needles.