Learn More
Seracare Life Sciences Inc AccuPlex SARS-CoV-2, Flu A/B & RSV Verification Panel, Full-process Control, Non-infectious Format, 4 x 3 mL
Supplier:  Seracare Life Sciences Inc 05050278
Encompass
The LGC Clinical Diagnostics AccuPlex™ SARS-CoV-2, Flu A/B and RSV Verification Panel is a non-infectious, molecular quality solution designed for multiplex tests. It significantly aids laboratories in assay verification, ensuring comprehensive performance documentation and proficiency evaluation. Utilizing innovative AccuPlex technology to mimic wild-type viruses, it stands out for its true viral protein coat, which provides robust full-process controls.
Catalog No. NC2267093