Learn More
Bioneer AccuPower Taq PCR PreMix, 0.2 ml thin-wall 8-tube strip with attached cap, 20 µl x 480 reactions
SDP
Catalog No. NC1586210
Encompass
The AccuPower Taq PCR PreMix is a convenient lyophilized PCR master mix containing Taq DNA polymerase, dNTPs, reaction buffer, tracking dye, and patented stabilizer and is aliquoted in 8-strip PCR tubes. The premix retains its activity for over a month at room temperature and is stable for two years in -20°C freezer.
Catalog No. NC1586210 Supplier Bioneer Supplier No. K2602
The AccuPower Taq PCR PreMix is a convenient lyophilized PCR master mix containing Taq DNA polymerase, dNTPs, reaction buffer, tracking dye, and patented stabilizer and is aliquoted in 8-strip PCR tubes. The premix retains its activity for over a month at room temperature and is stable for two years in -20°C freezer.