Learn More
VEOLIA WTS Accuracy/Precision Set, 500 mg/L
Catalog No. NC1127498
Encompass
Set includes: (1) Reagent Water Blank and (1) 500 μg C/L (as USP Sucrose) Standard Solution.
Catalog No. NC1127498 Supplier VEOLIA WTS Supplier No. STD 3101301
Set includes: (1) Reagent Water Blank and (1) 500 μg C/L (as USP Sucrose) Standard Solution.