Learn More
BD Cell Analysis 3P C6 Flow Cytometer 2-month Maintenance Kit
Supplier:  BD Cell Analysis 3P 653150
Encompass
Contains (2) Peristaltic Pump Tubing, (1) Sheath Bottle Filter, (2) Decon/Cleaner Bottle Filters and (1) In-Line Sheath Filter
Catalog No. NC0613888