Learn More
BD Accuri™ C6 Plus Flow Cytometer System
Catalog No. bdb661311 Shop All Waters Corporation Products
Change view
Click to view available options
Includes:
Accuri C6 Plus
Accuri C6 Plus, CSampler Plus
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
BDB661311 Accuri C6 Plus
BDB661310 Accuri C6 Plus, CSampler Plus
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. BDB661311 Supplier BD Supplier No. 661311
Add to Cart
Add to Cart
Request a Quote
This item is not returnable. View return policy
BD Accuri C6 Plus Flow Cytometer, with its compact footprint, light weight, and operational simplicity, supports a wide variety of applications including immunology, cell biology, plant and microbiology, and industrial applications.
  • The BD Accuri C6 Plus Flow Cytometer, with its compact 11 x 14.75 x 16.5 inch footprint, light weight of 30 lb and operational simplicity, supports a wide variety of applications including immunology, cell and cancer biology, plant and microbiology and industrial applications.
  • This portable and compact cell analyzer holds the analytical power of a cytometer twice its size.
  • Used in thousands of research studies, the BD Accuri C6 Plus System provides an easy-to-use option to your research toolbox.
TRUSTED_SUSTAINABILITY
Specifications
Product Line BD Accuri
Includes Accuri C6 Plus
WARNING: Reproductive Harm - www.P65Warnings.ca.gov