Learn More
Chemglass Life Sciences Compact Balance, 120 Grams, 115V
SDP
Catalog No. NC1844684
Encompass_Preferred
Accuris Compact balances have been designed to meet the basic needs of academic and research labs requiring weight determinations to increments of 0.01g to 0.1g. Their reliable and precise weighing technology utilizes gold-plated, ceramic capacitors to determine sample mass.
Catalog No. NC1844684 Supplier Chemglass Life Sciences Supplier No. CG794120
Accuris Compact balances have been designed to meet the basic needs of academic and research labs requiring weight determinations to increments of 0.01g to 0.1g. Their reliable and precise weighing technology utilizes gold-plated, ceramic capacitors to determine sample mass.