Learn More
Chemglass Life Sciences Compact Balance, 500 Grams, 115V
SDP
Supplier:  Chemglass Life Sciences CG794500
Encompass_Preferred
Compact Balance, 500 Grams, 115V. Accuris Compact balances have been designed to meet the basic needs of academic and research labs requiring weight determinations to increments of 0.01g to 0.1g. Their reliable and precise weighing technology utilizes gold-plated, ceramic capacitors to determine sample mass.7.5 x 11 inch footprint. The lower capacity units (120g, 300g, and 500g) are readable to 0.01 g, and come with a draft shield ring to prevent air currents from affecting measurements. These models feature a 4.5 inch diameter, stainless steel weigh pan. All balances in the Accuris Compact line offer large, brightly lit LCD panels, and are easy to operate using the intuitive and splash resistant control panel. Different modes allow weight to be displayed in various units: grams, ounces, carats, and simple programs can be used for piece counting, and percent deviation weighing.
Catalog No. NC1739016