Learn More
Alkali Scientific Accuris MyView? Compact UV TransilluminaAccuris MyView? Compact UV Transilluminatortor
SDP
Catalog No. 502336207
Encompass_Preferred
Introducing the SmartDoc 2.0. This complete system has been developed for safely viewing nucleic acids in agarose gels and also for capturing quality digital images with a smart phone camera. Blue light is used instead of UV, which is safer for lab personnel and will not damage DNA samples. The illumination base emits an intense blue light with a peak output of 460nm for optimum excitation of nucleic acids stained with green fluorescent dyes (SYBR Green, SmartGlow™, GelStar™, GelGreen™, etc). The orange cover fits on top of the illumination base and acts as a filter to absorb blue light, allowing visualization of the fluorescing samples. The cover can also be set at an angle for gel access and cutting out DNA bands for further study. Optional amber glasses are available (RE4000-VG SmartBlue Viewing Glasses). These are ideal for users that are doing a lot of band excision work, as they eliminate
Catalog No. 50-233-6207 Supplier Alkali Scientific Supplier No. RE3100
Introducing the SmartDoc 2.0. This complete system has been developed for safely viewing nucleic acids in agarose gels and also for capturing quality digital images with a smart phone camera. Blue light is used instead of UV, which is safer for lab personnel and will not damage DNA samples. The illumination base emits an intense blue light with a peak output of 460nm for optimum excitation of nucleic acids stained with green fluorescent dyes (SYBR Green, SmartGlow™, GelStar™, GelGreen™, etc). The orange cover fits on top of the illumination base and acts as a filter to absorb blue light, allowing visualization of the fluorescing samples. The cover can also be set at an angle for gel access and cutting out DNA bands for further study. Optional amber glasses are available (RE4000-VG SmartBlue Viewing Glasses). These are ideal for users that are doing a lot of band excision work, as they eliminate