Learn More
Braintree Scientific Accuris High Fidelity DNA Polymerase, 200 units
SDP
Catalog No. 501955548
Encompass
Accuris™ High Fidelity DNA Polymerase 50X higher fidelity than Taq DNA polymerase. Works with crude DNA samples. Ideal for cloning, mutagenesis and microarrays. Optimized buffer system with unique PCR enhancers. For applications requiring highly accurate amplification, choose Accuris High Fidelity DNA Polymerase. Modified for better sensitivity and higher activity for more reproducible performance, this polymerase will amplify a wide range of targets, including those that are GC or AT rich as well as crude samples. A 3-5 proofreading exonuclease activity and an error rate of 4.55x10-7 makes Accuris High Fidelity DNA Polymerase the perfect partner for cloning applications. The supplied 5X buffer with dNTPs is optimized for compatibility with a variety of targets. **Sold for research purposes only and not for human use**
Catalog No. 50-195-5548 Supplier Braintree Scientific Supplier No. PR1000HF200
Accuris™ High Fidelity DNA Polymerase 50X higher fidelity than Taq DNA polymerase. Works with crude DNA samples. Ideal for cloning, mutagenesis and microarrays. Optimized buffer system with unique PCR enhancers. For applications requiring highly accurate amplification, choose Accuris High Fidelity DNA Polymerase. Modified for better sensitivity and higher activity for more reproducible performance, this polymerase will amplify a wide range of targets, including those that are GC or AT rich as well as crude samples. A 3-5 proofreading exonuclease activity and an error rate of 4.55x10-7 makes Accuris High Fidelity DNA Polymerase the perfect partner for cloning applications. The supplied 5X buffer with dNTPs is optimized for compatibility with a variety of targets. **Sold for research purposes only and not for human use**