Learn More
Braintree Scientific Accuris Hot Start Master Mix, 2X Taq, 200 x 50ul Reactions
SDP
Catalog No. 501955557
Encompass
Hot Start Taq DNA Polymerase and Master Mix Exceptional sensitivity for low copy PCR Ideal for multiplex PCR and amplification of GC-rich DNA Same enhanced features as Ac Curis Taq Polymerase Buffer optimized for fast cycling and reproducibility Engineered for controlled polymerase activity, Accuris Hot Start Taq is bound with a monoclonal antibody that blocks enzyme activity. This allows reactions to be set up at room temperature without the risk of non-specific amplification. When samples are ready, the reaction mixture is heated to 95°C to denature the antibody and initiate the reaction. Similar to the standard Accuris Taq, Hot Start Taq is provided with a 5X buffer, or in a ready to use 2X Master Mix. The Master Mix can be ordered with or without a red gel loading dye. Researchers preferring a chemically modified
Catalog No. 50-195-5557 Supplier Braintree Scientific Supplier No. PR1001HS200
Hot Start Taq DNA Polymerase and Master Mix Exceptional sensitivity for low copy PCR Ideal for multiplex PCR and amplification of GC-rich DNA Same enhanced features as Ac Curis Taq Polymerase Buffer optimized for fast cycling and reproducibility Engineered for controlled polymerase activity, Accuris Hot Start Taq is bound with a monoclonal antibody that blocks enzyme activity. This allows reactions to be set up at room temperature without the risk of non-specific amplification. When samples are ready, the reaction mixture is heated to 95°C to denature the antibody and initiate the reaction. Similar to the standard Accuris Taq, Hot Start Taq is provided with a 5X buffer, or in a ready to use 2X Master Mix. The Master Mix can be ordered with or without a red gel loading dye. Researchers preferring a chemically modified