Learn More
Braintree Scientific Accuris Hot Start Master Mix, 2X Taq, 1000 x 50ul Reactions
SDP
Catalog No. 501955556
Encompass
Hot Start Taq DNA Polymerase and Master Mix Exceptional sensitivity for low copy PCR Ideal for multiplex PCR and amplification of GC-rich DNA Same enhanced features as Ac Curis Taq Polymerase Buffer optimized for fast cycling and reproducibility Engineered for controlled polymerase activity, Accuris Hot Start Taq is bound with a monoclonal antibody that blocks enzyme activity. This allows reactions to be set up at room temperature without the risk of non-specific amplification. When samples are ready, the reaction mixture is heated to 95C to denature the antibody and initiate the reaction. Similar to the standard Accuris Taq, Hot Start Taq is provided with a 5X buffer, or in a ready to use 2X Master Mix. The Master Mix can be ordered with or without a red gel loading dye. Researchers preferring a chemically modified **Sold for research purposes only and not for human use**
Catalog No. 50-195-5556 Supplier Braintree Scientific Supplier No. PR1001HS1000
Hot Start Taq DNA Polymerase and Master Mix Exceptional sensitivity for low copy PCR Ideal for multiplex PCR and amplification of GC-rich DNA Same enhanced features as Ac Curis Taq Polymerase Buffer optimized for fast cycling and reproducibility Engineered for controlled polymerase activity, Accuris Hot Start Taq is bound with a monoclonal antibody that blocks enzyme activity. This allows reactions to be set up at room temperature without the risk of non-specific amplification. When samples are ready, the reaction mixture is heated to 95C to denature the antibody and initiate the reaction. Similar to the standard Accuris Taq, Hot Start Taq is provided with a 5X buffer, or in a ready to use 2X Master Mix. The Master Mix can be ordered with or without a red gel loading dye. Researchers preferring a chemically modified **Sold for research purposes only and not for human use**