Learn More
Pyramex™ Accurist™ Safety Eyewear
Catalog No. 19155913 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Lens Tint:
Clear
Gray
Mirror, Silver
Silver
Frame Color:
Black
Black, Red
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint Frame Color
19-155-913 Clear Black, Red
19-155-857 Clear Black
19-155-858 Gray Black
19-155-859 Silver Black
19-155-860 Mirror, Silver Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-155-913 Supplier Pyramex™ Supplier No. SBR8710DTP
Add to Cart
Add to Cart
This item is not returnable. View return policy
Sporty design for on and off the job
  • Inner foam padding is flame resistant
  • Built-in rubber nosepiece
  • Durable frame features dual-injected straightback temples
  • Anti-fog lenses available
  • Scratch resistant polycarbonate lens provides 99% UVA/B/C protection
  • Specifications
    Lens Coating Anti-fog
    Certifications/Compliance ANSI Z87.1,CSA Z94.3-07
    Description Full eye protection 10 curve lens
    Lens Tint Clear
    Frame Color Black, Red
    Temple Style Spatula with double injection rubber
    Lens Material Polycarbonate
    Frame Material Nylon
    Frame Type Dual Lens with Side Protection