Learn More
Sigma Aldrich Fine Chemicals Biosciences AccurisTM Analytical Balan1EA
Catalog No. 502908809
Encompass
The Quick-Cal(TM) internal calibration feature allows quick and convenient calibration without the need for a calibration standard weight.
Catalog No. 50-290-8809 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7413561EA
The Quick-Cal(TM) internal calibration feature allows quick and convenient calibration without the need for a calibration standard weight.