Learn More
Sigma Aldrich Fine Chemicals Biosciences AccurisTM Analytical Balan1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z7413491EA
Encompass
The Quick-Cal(TM) internal calibration feature allows quick and convenient calibration without the need for a calibration standard weight.
Catalog No. 50-290-8561