Learn More
Sigma Aldrich Fine Chemicals Biosciences AccurisTM Compact Balance1EA
Catalog No. 502906169
Encompass
Accuris(TM) Compact balances have been designed to meet the basic needs of academic and research labs requiring weight determinations to increments of 0.01 g to 0.1 g. Their reliable and precise weighing technology utilizes gold-plated ceramic capacitors to determine sample mass.The Compact series includes 6 models covering a wide range of maximum weight capacities and come with a draft shield ring to prevent air currents from affecting measurements.The larger capacity models (1000 g 5000 g and 10 000 g) do not include the draft shield ring.
Catalog No. 50-290-6169 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7413841EA
Accuris(TM) Compact balances have been designed to meet the basic needs of academic and research labs requiring weight determinations to increments of 0.01 g to 0.1 g. Their reliable and precise weighing technology utilizes gold-plated ceramic capacitors to determine sample mass.The Compact series includes 6 models covering a wide range of maximum weight capacities and come with a draft shield ring to prevent air currents from affecting measurements.The larger capacity models (1000 g 5000 g and 10 000 g) do not include the draft shield ring.