Learn More
Sigma Aldrich Fine Chemicals Biosciences AccurisTM E2 Grade Calibra1EA
Catalog No. 502906023
Encompass
Premium Class E2 Weight Set is ideal for calibration of analytical balances with a 0.0001g readability. Precision manufactured from high purity 316 stainless steel these weights are corrosion resistant with magnetic susceptibility 0.005. The set includes 6 weights 100mg to 100g for checking repeatability cornerload linearity and span. The weights are supplied in a protective case with tweezers a cleaning cloth and a certificate of verification.
Catalog No. 50-290-6023 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7423111EA
Premium Class E2 Weight Set is ideal for calibration of analytical balances with a 0.0001g readability. Precision manufactured from high purity 316 stainless steel these weights are corrosion resistant with magnetic susceptibility 0.005. The set includes 6 weights 100mg to 100g for checking repeatability cornerload linearity and span. The weights are supplied in a protective case with tweezers a cleaning cloth and a certificate of verification.