Learn More
Sigma Aldrich Fine Chemicals Biosciences AccurisTM f1 Grade Calibra1EA
Catalog No. 502906106
Encompass
Precision compact and portable balances (readability 0.1 to 0.001g) can be calibrated with our Basic Class f1 Calibration Set. Included in the set are 23 stainless steel cylindrical and leaf weights 2 x 200g 1 x 100g 1 x 50g 2 x 20g 1 x 10g 1 x 5g 2 x 2g 1 x 1g 1 x 500mg 2 x 200mg 1 x 100mg 1 x 50mg 2 x 20mg 1 x 10mg 1 x 5mg 2 x 2mg 1 x 1mg. Cylindrical weights are marked with their nominal value. All are supplied in a protective case along with tweezers a cleaning cloth and dust brush.
Catalog No. 50-290-6106 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. BMSW11002001EA
Precision compact and portable balances (readability 0.1 to 0.001g) can be calibrated with our Basic Class f1 Calibration Set. Included in the set are 23 stainless steel cylindrical and leaf weights 2 x 200g 1 x 100g 1 x 50g 2 x 20g 1 x 10g 1 x 5g 2 x 2g 1 x 1g 1 x 500mg 2 x 200mg 1 x 100mg 1 x 50mg 2 x 20mg 1 x 10mg 1 x 5mg 2 x 2mg 1 x 1mg. Cylindrical weights are marked with their nominal value. All are supplied in a protective case along with tweezers a cleaning cloth and dust brush.