Learn More
Sigma Aldrich Fine Chemicals Biosciences AccurisTM Precision Balanc1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z7413631EA
Encompass
Accuris(TM) Precision balances quickly and accurately display sample measurements on a large backlit LCD screen. Utilizing highly precise gold plated ceramic capacitance sensors these balances tare and stabilize quickly and provide reliable weight measurements within 0.001 grams.Six models are available in the Precision Balance range covering a wide range of maximum capacity requirements. The smaller capacity models (120 g 320 g and 500 g) include a glass draft shield with 3 sliding doors for easy access the weighing chamber. The larger capacity models (1200 g 3200 g and 5000 g) feature a more traditional top-loading design and come with larger weigh pans.Leveling feet on the front are easy to access and a leveling bubble integrated into the control panel aids in the leveling process.
Catalog No. 50-290-8576