Learn More
Sigma Aldrich Fine Chemicals Biosciences AccurisTM SmartDocTM 2.0 I1EA
Catalog No. 502908937
Encompass
Introducing the SmartDoc 2.0. This complete system has been developed for safely viewing nucleic acids in agarose gels and also for capturing quality digital images with a smart phone camera. Blue light is used instead of UV which is safer for lab personnel and will not damage DNA samples. The illumination base emits an intense blue light with a peak output of 460nm for optimum excitation of nucleic acids stained with safe green dyes (SYBR Green SmartGlow GelStar GelGreen etc.) The orange cover fits on top of the illumination base and acts as a filter to absorb blue light allowing visualization of the fluorescing samples. The cover can also be set at an angle for gel access and cutting out DNA bands for further study. The SmartDoc imaging enclosure includes an orange photo filter for photography of the gels with a smartphone. An extending adapter can be inserted for full dimension imaging of gels up to 15X15 cm or removed for close up pictures. The enclosure can also be used dire
Catalog No. 50-290-8937 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7425911EA
Introducing the SmartDoc 2.0. This complete system has been developed for safely viewing nucleic acids in agarose gels and also for capturing quality digital images with a smart phone camera. Blue light is used instead of UV which is safer for lab personnel and will not damage DNA samples. The illumination base emits an intense blue light with a peak output of 460nm for optimum excitation of nucleic acids stained with safe green dyes (SYBR Green SmartGlow GelStar GelGreen etc.) The orange cover fits on top of the illumination base and acts as a filter to absorb blue light allowing visualization of the fluorescing samples. The cover can also be set at an angle for gel access and cutting out DNA bands for further study. The SmartDoc imaging enclosure includes an orange photo filter for photography of the gels with a smartphone. An extending adapter can be inserted for full dimension imaging of gels up to 15X15 cm or removed for close up pictures. The enclosure can also be used dire