Learn More
LAMDA BIOTECH INC AccuRuler Broad Protein Ladder
Catalog No. NC3724889
Encompass_Preferred
A pre-stained protein molecular weight marker containing 12 proteins with expanded range from 10 to 310 kDa. Designed for protein separation monitoring and molecular weight determination in electrophoresis and Western blot applications.
Catalog No. NC3724889 Supplier LAMDA BIOTECH INC Supplier No. G02103
A pre-stained protein molecular weight marker containing 12 proteins with expanded range from 10 to 310 kDa. Designed for protein separation monitoring and molecular weight determination in electrophoresis and Western blot applications.