Learn More
LAMDA BIOTECH INC AccuRuler + Prest Prot Ladder
Catalog No. NC3724883
Encompass_Preferred
A pre-stained protein molecular weight marker containing 12 proteins ranging from 10 to 245 kDa, formulated for protein electrophoresis and Western blot applications.
Catalog No. NC3724883 Supplier LAMDA BIOTECH INC Supplier No. G02102
A pre-stained protein molecular weight marker containing 12 proteins ranging from 10 to 245 kDa, formulated for protein electrophoresis and Western blot applications.