Learn More
LAMDA BIOTECH INC AccuRuler + Protein Ladder 10
Catalog No. NC3724886
Encompass_Preferred
The AccuRuler Plus™ Prestained Protein Ladder is an extended-range protein molecular weight marker containing 12 pre-stained proteins spanning 10-245kDa. This multi-tube format includes distinctive red (75kDa) and green (25kDa) reference bands for easy molecular weight determination.
Catalog No. NC3724886 Supplier LAMDA BIOTECH INC Supplier No. G0210210
The AccuRuler Plus™ Prestained Protein Ladder is an extended-range protein molecular weight marker containing 12 pre-stained proteins spanning 10-245kDa. This multi-tube format includes distinctive red (75kDa) and green (25kDa) reference bands for easy molecular weight determination.