Learn More
LAMDA BIOTECH INC AccuRuler Protein Ladder10 tb
Catalog No. NC3724882
Encompass_Preferred
The AccuRuler™ Prestained Protein Ladder is a precision protein molecular weight marker containing 10 pre-stained proteins spanning 10-180kDa. This multi-tube format includes distinctive red (75kDa) and green (25kDa) reference bands for easy molecular weight determination.
Catalog No. NC3724882 Supplier LAMDA BIOTECH INC Supplier No. G0210110
The AccuRuler™ Prestained Protein Ladder is a precision protein molecular weight marker containing 10 pre-stained proteins spanning 10-180kDa. This multi-tube format includes distinctive red (75kDa) and green (25kDa) reference bands for easy molecular weight determination.