Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SeraCare™ ACCURUN 1™ Multi-Analyte Independent Positive Controls

Catalog No. 22554503 Shop All SeraCare Life Sciences Products
Change view
Click to view available options
Certifications/Compliance:
CE, IVD, CE
CE, IVD, CE, HC
Type:
ACCURUN 1 Series 2400
ACCURUN 1 Series 2500
ACCURUN 1 Series 4000
ACCURUN 1 Series 8100
For Use With (Equipment):
Abbott Prism™
BioMerieux™ Vidas™ HIV DUO
Siemens Enzygnost™, HIV INTEGRA™
Quantity:
3 x 5 mL
12 x 3.5 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance Type For Use With (Equipment) Quantity
22-554-503 CE, IVD, CE ACCURUN 1 Series 4000 BioMerieux™ Vidas™ HIV DUO 3 x 5 mL
22-554-301 CE, IVD, CE, HC ACCURUN 1 Series 2500 Abbott Prism™ 12 x 3.5 mL
22-554-500 CE, IVD, CE, HC ACCURUN 1 Series 2400 Abbott Prism™ 12 x 3.5 mL
22-554-506 CE, IVD, CE ACCURUN 1 Series 8100 Siemens Enzygnost™, HIV INTEGRA™ 3 x 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy

Use the Centers for Disease Control (CDC) recommended universal precautions for handling ACCURUN 1 controls. Do not pipette by mouth; do not eat or drink in areas where specimens are being handled. Clean any spillage by immediately wiping up with 0.5% sodium hypochlorite solution. Dispose of all specimens, controls and materials used in testing as thought they contain infectious agents.

To prevent formation of potentially explosive compounds due to reactions of sodium azide and copper or lead pipes, flush waste lines with large quantities of water.

Catalog No. 22-554-503 Supplier SeraCare™ Supplier No. 20000008
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Use the Centers for Disease Control (CDC) recommended universal precautions for handling ACCURUN 1 controls. Do not pipette by mouth; do not eat or drink in areas where specimens are being handled. Clean any spillage by immediately wiping up with 0.5% sodium hypochlorite solution. Dispose of all specimens, controls and materials used in testing as thought they contain infectious agents.

To prevent formation of potentially explosive compounds due to reactions of sodium azide and copper or lead pipes, flush waste lines with large quantities of water.

Independent, multi-analyte control that helps with compliance of regulatory requirements

ACCURUN 1 controls are formulated for use with in vitro diagnostic test methods for the qualitative detection of HBsAg and antibodies or antigens to multiple bloodborne viruses

  • Multi-Analyte Positive Controls for Hepatitis (HBc, HCV, HBsAg), HIV (1 and 2), HTLV (I and II) and CMV
  • Helps to enhance quality control and quality assurance in testing labs
  • Human plasma based controls mimic human samples
  • Liquid, ready-to-use - no reconstitution or special handling required
  • Stable at refrigerated temperatures
  • Controls are specially formulated as low level positive for most manual and automated assay platforms
  • Independent controls meet compliance requirements of regulatory agencies and certifying organizations
  • Extended shelf life means efficiency for labs, allowing assay performance to be monitored with same lot over an extended period of time

  • SeraCare facilities are ISO 9001 and 13485 certified and comply with cGMP standards

Certifications

CE marked

Compliance

IVD, HC (22-554-500, 22-554-469, 22-554-470, 22-554-301, 21-123-102)

Specifications

Certifications/Compliance CE, IVD, CE
Content And Storage 2°C to 8°C
Format Vial
For Use With (Equipment) BioMerieux™ Vidas™ HIV DUO
Type ACCURUN 1 Series 4000
Intended Use Detection of Antibodies to HIV 1 and 2, Antibodies to HTLV I and II, Antibodies to HBcAg, Antibodies to HCV, Antibodies to Cytomegalovirus, and HBsAg
Quantity 3 x 5 mL
Source Human Serum or Plasma
Stability 60 Days Open Vial
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.