Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SeraCare™ ACCURUN™ HPV Positive and Negative Controls

Catalog No. 22554593 Shop All SeraCare Life Sciences Products
Change view
Click to view available options
Quantity:
10 x 0.8 mL Cell Supension, 10 x 2 mL Diluent
10 x 1 mL Cell Supension, 10 x 1 mL Diluent
20 x 4 mL
Type:
ACCURUN 372 HPV DNA Positive Control
ACCURUN 872 HPV DNA Negative Control
ACCURUN 873 HPV DNA Negative Control
Certifications/Compliance:
CE
IVD, CE, HC
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type Certifications/Compliance
22-554-593 10 x 0.8 mL Cell Supension, 10 x 2 mL Diluent ACCURUN 872 HPV DNA Negative Control CE
22-564-332 20 x 4 mL ACCURUN 873 HPV DNA Negative Control IVD, CE, HC
22-554-594 10 x 1 mL Cell Supension, 10 x 1 mL Diluent ACCURUN 372 HPV DNA Positive Control CE
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 22-554-593 Supplier SeraCare™ Supplier No. 20350009
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Independent positive and negative controls to enhance quality control and quality assurance in the diagnostic testing lab

  • Formulated for use with laboratory tests that detect Human Papilloma Virus (HPV) DNA in human cervical samples
  • For research use only: Not for use in diagnostic procedures

ACCURUN 372 Series

  • Manufactured from cultured human epithelial cells (SiHa) containing an integrated genome of HPV type 16
  • SiHa cells are mixed with noninfected cultured human cells and suspended in solution
  • Vial of diluent also supplied
ACCURUN 378 Series
  • Contains transfected cells with full-length episomal HPV16 DNA, the HPV type most often present in cervical carcinomas
  • Transfected cells are mixed with noninfected cultured human cells and preserved in solution
ACCURUN 872 and 873
  • Manufactured from cultured human cells and preserved in solution

Specifications

Certifications/Compliance CE
Type ACCURUN 872 HPV DNA Negative Control
Intended Use Detect Human Papilloma Virus DNA in Human Cervical samples
Quantity 10 x 0.8 mL Cell Supension, 10 x 2 mL Diluent
Source Human Cells
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.