Learn More
SeraCare™ ACCURUN™ HPV Positive and Negative Controls
Catalog No. 22564332 Shop All SeraCare Life Sciences Products
Change view
Click to view available options
Quantity:
10 x 0.8 mL Cell Supension, 10 x 2 mL Diluent
20 x 4 mL
Type:
ACCURUN 872 HPV DNA Negative Control
ACCURUN 873 HPV DNA Negative Control
Certifications/Compliance:
CE
IVD, CE, HC
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type Certifications/Compliance
22-564-332 20 x 4 mL ACCURUN 873 HPV DNA Negative Control IVD, CE, HC
22-554-593 10 x 0.8 mL Cell Supension, 10 x 2 mL Diluent ACCURUN 872 HPV DNA Negative Control CE
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-564-332 Supplier SeraCare™ Supplier No. 20350005
Add to Cart
Add to Cart
Independent positive and negative controls to enhance quality control and quality assurance in the diagnostic testing lab
  • Formulated for use with laboratory tests that detect Human Papilloma Virus (HPV) DNA in human cervical samples
  • For research use only: Not for use in diagnostic procedures
ACCURUN 372 Series
  • Manufactured from cultured human epithelial cells (SiHa) containing an integrated genome of HPV type 16
  • SiHa cells are mixed with noninfected cultured human cells and suspended in solution
  • Vial of diluent also supplied
ACCURUN 378 Series
  • Contains transfected cells with full-length episomal HPV16 DNA, the HPV type most often present in cervical carcinomas
  • Transfected cells are mixed with noninfected cultured human cells and preserved in solution
ACCURUN 872 and 873
  • Manufactured from cultured human cells and preserved in solution
Specifications
Certifications/Compliance IVD, CE, HC
Type ACCURUN 873 HPV DNA Negative Control
Intended Use Detect Human Papilloma Virus DNA in Human Cervical samples
Quantity 20 x 4 mL