Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SeraCare™ ACCURUN™ QC Serology Single Analyte Positive Controls

Catalog No. 22554175 Shop All SeraCare Life Sciences Products
Change view
Click to view available options
Quantity:
1 x 1 mL
3 x 1 mL
Type:
ACCURUN 130 Lyme IgG Positive Control
ACCURUN 132 Lyme IgM Positive Control
ACCURUN 140 Rubella IgG Positive Control
ACCURUN 145 CMV IgG Positive Control
ACCURUN 156 Reagin (Syphilis) Positive Control
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type
22-554-175 1 x 1 mL ACCURUN 130 Lyme IgG Positive Control
22-554-182 1 x 1 mL ACCURUN 145 CMV IgG Positive Control
22-554-187 3 x 1 mL ACCURUN 156 Reagin (Syphilis) Positive Control
22-554-179 1 x 1 mL ACCURUN 140 Rubella IgG Positive Control
22-554-176 1 x 1 mL ACCURUN 132 Lyme IgM Positive Control
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 22-554-175 Supplier SeraCare™ Supplier No. 20150080
Add to Cart
Add to Cart

Offer the ultimate in flexibility and convenience

Quality control for infectious disease testing laboratories.

  • Detect and prevent errors
  • Monitor assay performance
  • Simplify testing procedures
  • Document regulatory compliance
  • Contain a single antibody or antigen and are designed to perform in the dynamic range of most manufacturers' assays
  • Designed to mimic a patient sample
  • Supplied ready to use, requiring no reconstitution or special handling

Specifications

Certifications/Compliance IVD, CE
Content And Storage 2°C to 8°C
Format Vial
For Use With (Equipment) BioMerieux™ Vidas™
Type ACCURUN 130 Lyme IgG Positive Control
Intended Use Qualitative detection of IgG Antibodies to Borrelia burgdorferi (Lyme disease)
Quantity 1 x 1 mL
Source Human Serum or Plasma
Stability 60 Days Open Vial
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.