Learn More
World Precision Instrument Accuscope Binocular Microscope, Plan Achromat
SDP
Catalog No. 502539387
Encompass
ACCU-SCOPE's 3012 Series features E-Plan Achromat Infinity objectives which are renowned for crisp, high contrast images with outstanding resolution. Modular in design, the 3012 boasts options for entry level fluorescence applications, phase contrast, darkfield and simple polarization. With 22mm field of view eyepieces and an eyepoint height that is adjustable, it can be raised 35mm by rotating the eyepiece tubes up 180° to accommodate the needs of various users. ACCU-SCOPE's Infinity E-plan objectives provide users with best in class high resolution, high contrast objectives. Objectives are available in plan achromat and plan phase configurations. The rackless, scratch resistant rectangular stage has low-positioned X-Y coaxial controls. A clinical level microscope for routine applications in medical, veterinary and university labs.
Catalog No. 50-253-9387 Supplier World Precision Instrument Supplier No. 504220
ACCU-SCOPE's 3012 Series features E-Plan Achromat Infinity objectives which are renowned for crisp, high contrast images with outstanding resolution. Modular in design, the 3012 boasts options for entry level fluorescence applications, phase contrast, darkfield and simple polarization. With 22mm field of view eyepieces and an eyepoint height that is adjustable, it can be raised 35mm by rotating the eyepiece tubes up 180° to accommodate the needs of various users. ACCU-SCOPE's Infinity E-plan objectives provide users with best in class high resolution, high contrast objectives. Objectives are available in plan achromat and plan phase configurations. The rackless, scratch resistant rectangular stage has low-positioned X-Y coaxial controls. A clinical level microscope for routine applications in medical, veterinary and university labs.