Learn More
World Precision Instrument Accuscope Trinocular Microscope, Infinity Plan achromat
SDP
Catalog No. 502539375
Encompass
The ACCU-SCOPE 3000-LED Microscope Series delivers outstanding optical performance, value and resolution to meet the exacting standards of life science professionals and students. With a newly designed infinity plan optical system, a best-in-class 20 mm field of view and a super-bright 3-watt LED illuminator, the 3000-LED Series provides high contrast images with outstanding resolution, precision design and enhanced illumination.
Catalog No. 50-253-9375 Supplier World Precision Instrument Supplier No. 504443
The ACCU-SCOPE 3000-LED Microscope Series delivers outstanding optical performance, value and resolution to meet the exacting standards of life science professionals and students. With a newly designed infinity plan optical system, a best-in-class 20 mm field of view and a super-bright 3-watt LED illuminator, the 3000-LED Series provides high contrast images with outstanding resolution, precision design and enhanced illumination.