Learn More
Sigma Aldrich Fine Chemicals Biosciences ACCUSPINTM System Histopaq
Catalog No. 502905355
Encompass
gamma-irradiated 12 mL polypropylene tubes fitted with a HDPE porous barrier and aseptically filled with 3 mL of Histopaque 1077. Each tube will separate 3-6 mL of anticoagulated blood.
Catalog No. 50-290-5355 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A692940X3ML
gamma-irradiated 12 mL polypropylene tubes fitted with a HDPE porous barrier and aseptically filled with 3 mL of Histopaque 1077. Each tube will separate 3-6 mL of anticoagulated blood.