Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AccuStandard, Inc ACCUSTANDARD Bisphenol BP, Certified Reference Material, 10 mg/mL, in Methanol
SDP

Catalog No. NC3991826
Encompass

This product is a Certified Reference Material (CRM) of Bisphenol BP, provided as a 10 mg/mL solution in Methanol. It is a single component standard with a molecular formula of C25H20O2 and a molecular weight of 352.43.

  • Certified reference material for accurate analysis.
  • Single component standard.
  • Supplied at a concentration of 10 mg/mL in methanol.
  • Molecular formula of C25H20O2.
  • Molecular weight of 352.43.
  • Melting point of 125°C.
  • Flash point greater than 200°C.
  • Recommended storage condition is ambient (>5°C).

Catalog No. NC3991826 Supplier AccuStandard, Inc Supplier No. BPABPS
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product is a Certified Reference Material (CRM) of Bisphenol BP, provided as a 10 mg/mL solution in Methanol. It is a single component standard with a molecular formula of C25H20O2 and a molecular weight of 352.43.

  • Certified reference material for accurate analysis.
  • Single component standard.
  • Supplied at a concentration of 10 mg/mL in methanol.
  • Molecular formula of C25H20O2.
  • Molecular weight of 352.43.
  • Melting point of 125°C.
  • Flash point greater than 200°C.
  • Recommended storage condition is ambient (>5°C).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.