Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AccuStandard, Inc AccuStandard Certified Reference Material, Bisphenol M, 10 mg/mL, Methanol, 1 mL
SDP

Catalog No. NC3991852
Encompass

This product is a Certified Reference Material for Bisphenol M. It is provided at a target concentration of 10 mg/mL in Methanol, with a total volume of 1 mL. It is intended for analytical and scientific applications requiring high accuracy and traceability.

  • Certified Reference Material classification
  • Target concentration of 10 mg/mL in Methanol
  • Single component formulation
  • Storage at ambient conditions (>5 °C)
  • CAS number: 13595-25-0
  • Volume: 1 mL

Catalog No. NC3991852 Supplier AccuStandard, Inc Supplier No. BPAMS
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product is a Certified Reference Material for Bisphenol M. It is provided at a target concentration of 10 mg/mL in Methanol, with a total volume of 1 mL. It is intended for analytical and scientific applications requiring high accuracy and traceability.

  • Certified Reference Material classification
  • Target concentration of 10 mg/mL in Methanol
  • Single component formulation
  • Storage at ambient conditions (>5 °C)
  • CAS number: 13595-25-0
  • Volume: 1 mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.