Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AccuStandard, Inc AccuStandard Certified Reference Material, Bisphenol P, 10 mg/mL, in Methanol
SDP

Catalog No. NC3991849
Encompass

This certified reference material contains Bisphenol P at a target concentration of 10 mg/mL in Methanol.

  • Contains Bisphenol P analyte.
  • Target concentration of 10 mg/mL.
  • Supplied in Methanol matrix.
  • Unit volume is 1 mL.
  • Recommended storage condition: Ambient (>5 °C).

Catalog No. NC3991849 Supplier AccuStandard, Inc Supplier No. BPAPS
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This certified reference material contains Bisphenol P at a target concentration of 10 mg/mL in Methanol.

  • Contains Bisphenol P analyte.
  • Target concentration of 10 mg/mL.
  • Supplied in Methanol matrix.
  • Unit volume is 1 mL.
  • Recommended storage condition: Ambient (>5 °C).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.