Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AccuStandard, Inc AccuStandard Certified Reference Material, Bisphenol PH, 10 mg/mL in Methanol, 1 mL
SDP

Catalog No. NC3991845
Encompass

This Certified Reference Material (CRM) from AccuStandard contains Bisphenol PH at a concentration of 10 mg/mL in Methanol. It is suitable for various analytical applications requiring a high-quality reference standard for Bisphenol PH (CAS 24038-68-4).

  • Contains bisphenol PH at a concentration of 10 mg/mL
  • Supplied in methanol solvent
  • Unit size of 1 mL
  • Recommended storage condition is ambient (>5 °C)
  • Melting point is -93.9 °C
  • Boiling point is 65 °C
  • Flash point is 52 °F (11 °C) (tcc)

Catalog No. NC3991845 Supplier AccuStandard, Inc Supplier No. BPAPHS
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This Certified Reference Material (CRM) from AccuStandard contains Bisphenol PH at a concentration of 10 mg/mL in Methanol. It is suitable for various analytical applications requiring a high-quality reference standard for Bisphenol PH (CAS 24038-68-4).

  • Contains bisphenol PH at a concentration of 10 mg/mL
  • Supplied in methanol solvent
  • Unit size of 1 mL
  • Recommended storage condition is ambient (>5 °C)
  • Melting point is -93.9 °C
  • Boiling point is 65 °C
  • Flash point is 52 °F (11 °C) (tcc)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.