Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AccuStandard, Inc AccuStandard Certified Reference Material, Bisphenol S, 10 mg/mL, 1 mL
SDP

Catalog No. NC3991846
Encompass

This product is a Certified Reference Material (CRM) from AccuStandard, featuring Bisphenol S at a concentration of 10 mg/mL in methanol. It is provided in a 1 mL unit, suitable for various laboratory applications requiring a reliable standard.

  • Certified reference material
  • Contains Bisphenol S
  • Target concentration: 10 mg/mL
  • Methanol as solvent
  • Unit volume: 1 mL
  • Ambient storage condition (above 5 °C)
  • Melting point: -93.9 °C
  • Boiling point: 65 °C
  • Flash point: 52 °F (11 °C)

Catalog No. NC3991846 Supplier AccuStandard, Inc Supplier No. BPASS
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product is a Certified Reference Material (CRM) from AccuStandard, featuring Bisphenol S at a concentration of 10 mg/mL in methanol. It is provided in a 1 mL unit, suitable for various laboratory applications requiring a reliable standard.

  • Certified reference material
  • Contains Bisphenol S
  • Target concentration: 10 mg/mL
  • Methanol as solvent
  • Unit volume: 1 mL
  • Ambient storage condition (above 5 °C)
  • Melting point: -93.9 °C
  • Boiling point: 65 °C
  • Flash point: 52 °F (11 °C)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.