Learn More
AccuStandard, Inc AccuStandard Certified Reference Standard, Bisphenol C, 10 mg/mL, in Methanol
SDP
Catalog No. NC3991827
Encompass
AccuStandard offers certified reference standards for bisphenol analogs, which are chemicals structurally similar to BPA and used as alternatives in plastics and resins. This specific reference standard is Bisphenol C, provided as a single component in methanol.
  • Bisphenol C reference standard
  • 10 mg/mL concentration in methanol
  • Single component formulation
  • CAS number: 79-97-0
  • Supplied in a 1 mL unit size
Catalog No. NC3991827 Supplier AccuStandard, Inc Supplier No. BPACS
AccuStandard offers certified reference standards for bisphenol analogs, which are chemicals structurally similar to BPA and used as alternatives in plastics and resins. This specific reference standard is Bisphenol C, provided as a single component in methanol.
  • Bisphenol C reference standard
  • 10 mg/mL concentration in methanol
  • Single component formulation
  • CAS number: 79-97-0
  • Supplied in a 1 mL unit size