Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AccuStandard, Inc AccuStandard Certified Reference Standard, Bisphenol Z, 10 mg/mL, in Methanol
SDP

Catalog No. NC3991848
Encompass

This certified reference material contains Bisphenol Z at a concentration of 10 mg/mL, prepared in a methanol matrix. It is suitable for various laboratory applications requiring a reliable reference standard.

  • Classified as a certified reference material.
  • Contains a single component.
  • Recommended storage condition is ambient, above 5 °C.
  • Supplied in a 1 mL unit.
  • Melting point is -93.9 °C.
  • Boiling point is 65 °C.
  • Flash point is 52 °F (11 °C) (tcc).
  • Presents hazards including high flammability, organ damage, and toxicity if absorbed through skin, swallowed, or inhaled.
  • Causes irritation to skin and eyes.

Catalog No. NC3991848 Supplier AccuStandard, Inc Supplier No. BPAZS
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This certified reference material contains Bisphenol Z at a concentration of 10 mg/mL, prepared in a methanol matrix. It is suitable for various laboratory applications requiring a reliable reference standard.

  • Classified as a certified reference material.
  • Contains a single component.
  • Recommended storage condition is ambient, above 5 °C.
  • Supplied in a 1 mL unit.
  • Melting point is -93.9 °C.
  • Boiling point is 65 °C.
  • Flash point is 52 °F (11 °C) (tcc).
  • Presents hazards including high flammability, organ damage, and toxicity if absorbed through skin, swallowed, or inhaled.
  • Causes irritation to skin and eyes.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.