Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AccuStandard, Inc AccuStandard Diisononyl Phthalate (DINP), Tech Mix, 100 mg, Reference Material
SDP

Catalog No. NC3991857
Encompass

Diisononyl phthalate, also known as DINP, is a technical mix certified reference material. This analytical standard is provided in a 100 mg unit and is characterized by its specific chemical and physical properties, making it suitable for various analytical applications.

  • CAS number is 28553-12-0.
  • Supplied in a 100 mg unit.
  • Molecular formula is C26H42O4.
  • Molecular weight is 418.61.
  • Melting point ranges from -48°C to -54°F.
  • Boiling point ranges from 331°C to 628°F.
  • Flash point is 236°C to 457°F, closed cup.
  • Classified as a reference material under 17034.
  • Recommended storage condition is ambient (>5°C).

Catalog No. NC3991857 Supplier AccuStandard, Inc Supplier No. ALR102N
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Diisononyl phthalate, also known as DINP, is a technical mix certified reference material. This analytical standard is provided in a 100 mg unit and is characterized by its specific chemical and physical properties, making it suitable for various analytical applications.

  • CAS number is 28553-12-0.
  • Supplied in a 100 mg unit.
  • Molecular formula is C26H42O4.
  • Molecular weight is 418.61.
  • Melting point ranges from -48°C to -54°F.
  • Boiling point ranges from 331°C to 628°F.
  • Flash point is 236°C to 457°F, closed cup.
  • Classified as a reference material under 17034.
  • Recommended storage condition is ambient (>5°C).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.