Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AccuStandard, Inc AccuStandard Diisopropyl phthalate, Certified Reference Material, CAS 605-45-8
SDP

Catalog No. NC3991859
Encompass

This product is a Diisopropyl phthalate Certified Reference Material, ideal for analytical applications requiring high accuracy. It features a CAS number of 605-45-8 and a molecular formula of C14H18O4, with a molecular weight of 250.29.

  • Certified Reference Material (17034 Classification)
  • CAS number: 605-45-8
  • Molecular formula: C14H18O4
  • Molecular weight: 250.29
  • Melting point: 20°C (68°F)
  • Store at ambient conditions (>5°C)

Catalog No. NC3991859 Supplier AccuStandard, Inc Supplier No. PHTH019N
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product is a Diisopropyl phthalate Certified Reference Material, ideal for analytical applications requiring high accuracy. It features a CAS number of 605-45-8 and a molecular formula of C14H18O4, with a molecular weight of 250.29.

  • Certified Reference Material (17034 Classification)
  • CAS number: 605-45-8
  • Molecular formula: C14H18O4
  • Molecular weight: 250.29
  • Melting point: 20°C (68°F)
  • Store at ambient conditions (>5°C)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.