Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AccuStandard, Inc ACCUSTANDARD INC Certified Reference Material, Bisphenol B, 10 mg/mL, Methanol
SDP

Catalog No. NC3991825
Encompass

Bisphenol B is a Certified Reference Material provided as a 10 mg/mL solution in Methanol, suitable for various laboratory applications.

  • Provided as a 10 mg/mL solution in methanol.
  • Classified as a certified reference material.
  • Contains a single component for reliable analysis.
  • Can be stored at ambient temperatures (above 5 °C).
  • Features a melting point of -93.9 °C.
  • Boiling point is 65 °C.
  • Flash point is 52 °F (11 °C) (tcc).

Catalog No. NC3991825 Supplier AccuStandard, Inc Supplier No. BPABS
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bisphenol B is a Certified Reference Material provided as a 10 mg/mL solution in Methanol, suitable for various laboratory applications.

  • Provided as a 10 mg/mL solution in methanol.
  • Classified as a certified reference material.
  • Contains a single component for reliable analysis.
  • Can be stored at ambient temperatures (above 5 °C).
  • Features a melting point of -93.9 °C.
  • Boiling point is 65 °C.
  • Flash point is 52 °F (11 °C) (tcc).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.