Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AccuStandard, Inc ACCUSTANDARD INC Certified Reference Material, Bisphenol G, 10 mg/mL, Methanol
SDP

Catalog No. NC3991830
Encompass

Bisphenol G is a Certified Reference Material with a concentration of 10 mg/mL in Methanol. It is a highly flammable substance that causes damage to organs, and is toxic if absorbed through skin or swallowed. It is also harmful if inhaled and irritating to skin and eyes.

  • Certified reference material
  • Concentration of 10 mg/mL
  • Solvent is methanol
  • Unit size is 1 mL
  • Melting point is -93.9°C
  • Boiling point is 65°C
  • Flash point is 52°F (11°C) (tcc)
  • Highly flammable
  • Causes damage to organs
  • Toxic if absorbed through skin
  • Toxic if swallowed
  • Harmful if inhaled
  • Irritating to skin
  • Irritating to eyes

Catalog No. NC3991830 Supplier AccuStandard, Inc Supplier No. BPAGS
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bisphenol G is a Certified Reference Material with a concentration of 10 mg/mL in Methanol. It is a highly flammable substance that causes damage to organs, and is toxic if absorbed through skin or swallowed. It is also harmful if inhaled and irritating to skin and eyes.

  • Certified reference material
  • Concentration of 10 mg/mL
  • Solvent is methanol
  • Unit size is 1 mL
  • Melting point is -93.9°C
  • Boiling point is 65°C
  • Flash point is 52°F (11°C) (tcc)
  • Highly flammable
  • Causes damage to organs
  • Toxic if absorbed through skin
  • Toxic if swallowed
  • Harmful if inhaled
  • Irritating to skin
  • Irritating to eyes

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.