Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AccuStandard, Inc AccuStandard Reference Standard, Bisphenol C-dichloride, 10 mg/mL, in Methanol
SDP

Catalog No. NC3991853
Encompass

This certified reference material is a solution of Bisphenol C-dichloride at a concentration of 10 mg/mL in methanol. It is supplied in a 1 mL unit for analytical applications requiring precise and reliable standards.

  • Supplied as a 1 mL unit.
  • Concentration of 10 mg/mL of Bisphenol C-dichloride.
  • Solvent is methanol.
  • Classified as a certified reference material.
  • Storage condition is ambient (>5 °C).

Catalog No. NC3991853 Supplier AccuStandard, Inc Supplier No. BPAC2S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This certified reference material is a solution of Bisphenol C-dichloride at a concentration of 10 mg/mL in methanol. It is supplied in a 1 mL unit for analytical applications requiring precise and reliable standards.

  • Supplied as a 1 mL unit.
  • Concentration of 10 mg/mL of Bisphenol C-dichloride.
  • Solvent is methanol.
  • Classified as a certified reference material.
  • Storage condition is ambient (>5 °C).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.