Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AccuStandard, Inc AccuStandard Reference Standard, Diisooctyl Phthalate, 100%
SDP

Supplier:  AccuStandard, Inc ALR103N

Encompass

This product is a Diisooctyl phthalate reference standard, provided as a clear, colorless, viscous liquid for laboratory applications. It is a stable chemical with a concentration of 100%.

  • Diisooctyl phthalate composition
  • 100% concentration
  • Clear, colorless, viscous liquid appearance
  • Suitable for laboratory chemical reference material applications
  • Molecular formula C24H38O4
  • Molecular weight 390.56
  • Melting point -50°C
  • Boiling point 235°C
  • Flash point 227°C (closed cup)
  • Specific gravity 0.98 g/cm3
  • Solubility in water 0.09 mg/L
  • Autoignition temperature 390°C
  • Stable chemical

Catalog No. NC3991856


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.