Learn More
STEMCELL Technologies ACCUTASE™, Size: 100 mL
SDP
Catalog No. NC1793126
Encompass
A cell detachment solution of proteolytic and collagenolytic enzymes. Useful for the routine detachment of cells from standard tissue culture plasticware and adhesion coated plasticware. ACCUTASE™ does not contain mammalian or bacterial-derived products. Each lot of ACCUTASE™ is tested for sterility (by USP membrane filtration method), enzymatic activity (tested with synthetic chromagenic tetrapeptides) and cell detachment from tissue culture plastic.
Catalog No. NC1793126 Supplier STEMCELL Technologies Supplier No. 07920
A cell detachment solution of proteolytic and collagenolytic enzymes. Useful for the routine detachment of cells from standard tissue culture plasticware and adhesion coated plasticware. ACCUTASE™ does not contain mammalian or bacterial-derived products. Each lot of ACCUTASE™ is tested for sterility (by USP membrane filtration method), enzymatic activity (tested with synthetic chromagenic tetrapeptides) and cell detachment from tissue culture plastic.