Learn More
Innovative Cell Technologies ACCUTASE CELL DETACHMENT REAGN
SDP
Catalog No. NC9839010
Encompass
Accutase; Sterile; 100mL Enzyme cell detachment medium; Composition: Proprietary formulation of enzymes, water, phosphate buffered saline, phenol red; Stability: Stable under normal temperatures and pressures
Catalog No. NC9839010 Supplier Innovative Cell Technologies Supplier No. AT104
Accutase; Sterile; 100mL Enzyme cell detachment medium; Composition: Proprietary formulation of enzymes, water, phosphate buffered saline, phenol red; Stability: Stable under normal temperatures and pressures